siRNA Details
Gene Symbol: 1810053B23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040486.1 | 4 | 673 | |||
NR_040488.1 | 4 | 515 | ||||
NR_040489.1 | 4 | 513 | ||||
GenBank mRNA | AI390631.1 | - | 4 | 505 | ||
AK007854.1 | - | 4 | 674 | |||
AK160158.1 | - | 4 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707857.1 | - | 4 | 674 | Chr. 16 - 93344020 - 93353189 [-] on Build GRCm38 |
