siRNA Details
Gene Symbol: 1810053B23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040486.1 | 4 | 717 | |||
NR_040488.1 | 4 | 559 | ||||
NR_040489.1 | 4 | 557 | ||||
GenBank mRNA | AK007854.1 | - | 4 | 718 | ||
AK160158.1 | - | 4 | 560 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707857.1 | - | 4 | 718 | Chr. 16 - 93344020 - 93353189 [-] on Build GRCm38 |
