siRNA Details
Gene Symbol: Gm38793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085647.1 | - | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661660.1 | - | 4 | 509 | Chr. 6 - 38918746 - 38984325 [-] on Build GRCm38 |
