siRNA Details
Gene Symbol: Gm20756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040771.1 | 3 | 291 | |||
GenBank mRNA | AK165854.1 | - | 3 | 292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY733948.1 | - | 3 | 292 | Chr. 6 - 25931426 - 25943142 [+] on Build GRCm38 |
