siRNA Details
Gene Symbol: Gm39321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136151.1 | - | 623 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720100.1 | - | 3 | 623 | Chr. 9 - 42142087 - 42145134 [+] on Build GRCm38 |
