siRNA Details
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA671314.1 | - | 1 | 144 | ||
AK017596.1 | - | 1 | 233 | |||
AK156477.1 | - | 1 | 1146 | |||
AK158707.1 | - | 1 | 1082 | |||
BE981800.1 | - | 1 | 454 |
