siRNA Details
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA671314.1 | - | 1 | 257 | ||
AK017596.1 | - | 1 | 346 | |||
AK156477.1 | - | 1, 2 | 1259 | |||
AK158707.1 | - | 1 | 1195 | |||
BE981800.1 | - | 1 | 567 |
