siRNA Details
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA671314.1 | - | 1 | 60 | ||
AK017596.1 | - | 1 | 149 | |||
AK156477.1 | - | 1 | 1062 | |||
AK158707.1 | - | 1 | 998 | |||
BE981800.1 | - | 1 | 370 |
