siRNA Details
Gene Symbol: 5430402P08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017263.1 | - | 2 | 549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717068.1 | - | 2 | 549 | Chr. 7 - 111366298 - 111395365 [+] on Build GRCm38 |
