siRNA Details
Gene Symbol: 9430031J08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020451.1 | - | 1 | 432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721697.1 | - | 1 | 432 | Chr. 15 - 33557052 - 33557665 [+] on Build GRCm38 |
