siRNA Details
Gene Symbol: 9430031K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020452.1 | - | 1 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721698.1 | - | 1 | 246 | Chr. 12 - 98762051 - 98762657 [+] on Build GRCm38 |
