siRNA Details
Gene Symbol: 4930579M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016330.1 | - | 1 | 292 | ||
AK131783.1 | - | 1 | 334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715969.1 | - | 1 | 292 | Chr. 8 - 47026885 - 47038177 [-] on Build GRCm38 |
