siRNA Details
Gene Symbol: Gm16619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079684.1 | - | 3 | 858 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB199376.2 | - | 1 | 350 | Chr. 16 - 49804202 - 49804874 [+] on Build GRCm38 |
