siRNA Details
Gene Symbol: 4933412E12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084517.1 | - | 3 | 639 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB475029.2 | - | 3 | 592 | Chr. 10 - 116950622 - 116959300 [+] on Build GRCm38 |
