siRNA Details
Gene Symbol: AI256396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046686.1 | - | 2609 | |||
AK134520.1 | - | 1544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX512726.1 | - | 1 | 507 | Chr. 6 - 100809553 - 100810075 [+] on Build GRCm38 |
