siRNA Details
Gene Symbol: C030007I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021055.1 | - | 1 | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728118.1 | - | 1 | 120 | Chr. 5 - 130844507 - 130844900 [+] on Build GRCm38 |
