siRNA Details
Gene Symbol: A930030B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020907.1 | - | 1 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742284.1 | - | 1 | 458 | Chr. 7 - 99901615 - 99902263 [-] on Build GRCm38 |
