siRNA Details
Gene Symbol: 1700061D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006849.1 | - | 1 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706830.1 | - | 1 | 386 | Chr. 5 - 105866373 - 105866775 [-] on Build GRCm38 |
