siRNA Details
Gene Symbol: 1700061A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006847.1 | - | 1 | 90 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706828.1 | - | 1 | 89 | Chr. 19 - 5488744 - 5489202 [-] on Build GRCm38 |
