siRNA Details
Gene Symbol: 1700060J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006843.1 | - | 716 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706824.1 | - | 2 | 716 | Chr. 4 - 53563067 - 53564697 [-] on Build GRCm38 |
