siRNA Details
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009110.1 | - | 1 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708950.1 | - | 1 | 367 | Chr. 2 - 101678465 - 101679125 [+] on Build GRCm38 |
