siRNA Details
Gene Symbol: 1600015H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005466.1 | - | 1 | 217 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705668.1 | - | 1 | 217 | Chr. 11 - 109344286 - 109344633 [+] on Build GRCm38 |
