siRNA Details
Gene Symbol: Gm12159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045100.1 | 3 | 1210 | |||
GenBank mRNA | AK041275.1 | - | 3 | 1211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB226319.2 | - | 1 | 471 | Chr. 11 - 45102872 - 45103574 [-] on Build GRCm38 |
