siRNA Details
Gene Symbol: Itprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014762.1 | - | 1 | 1271 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK626294.1 | - | 1 | 367 | Chr. 19 - 47917793 - 47918298 [-] on Build GRCm38 |
