siRNA Details
Gene Symbol: 2010015B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008245.1 | - | 1 | 285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV517683.1 | - | 1 | 386 | Chr. 12 - 83897861 - 83898322 [-] on Build GRCm38 |
