siRNA Details
Gene Symbol: Gm13112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030206.1 | - | 5 | 785 | ||
AK046472.1 | - | 1 | 3102 | |||
AV281208.2 | - | 4 | 593 | |||
BB321076.2 | - | 1 | 232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030206.1 | - | 5 | 785 | ||
AK046472.1 | - | 1 | 3102 | |||
AV281208.2 | - | 4 | 593 | |||
BB321076.2 | - | 1 | 232 |