siRNA Details
Gene Symbol: C530044C16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043004.1 | - | 5 | 1181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB251594.2 | - | 1 | 497 | Chr. 6 - 50814339 - 50814893 [+] on Build GRCm38 |
