siRNA Details
Gene Symbol: Lyrm7os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040284.1 | 3 | 907 | |||
GenBank mRNA | AK087691.1 | - | 3 | 908 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB553409.2 | - | 2 | 662 | Chr. 11 - 54821328 - 54824967 [+] on Build GRCm38 |
