siRNA Details
Gene Symbol: Arglu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034581.1 | - | 1439 | |||
AK038412.1 | - | 1077 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB087577.2 | - | 1 | 36 | Chr. 8 - 8665080 - 8665732 [-] on Build GRCm38 |
