siRNA Details
Gene Symbol: 9330182O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034361.1 | - | 2 | 230 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626390.1 | - | 2 | 230 | Chr. 15 - 40134365 - 40148581 [+] on Build GRCm38 |
