siRNA Details
Gene Symbol: 4930527G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045395.1 | 4 | 490 | |||
GenBank mRNA | AK019704.1 | - | 4 | 491 | ||
BY715476.1 | - | 4 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV267612.1 | - | 2 | 181 | Chr. 18 - 29452123 - 29466107 [+] on Build GRCm38 |
