siRNA Details
Gene Symbol: 4930527G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045395.1 | 3, 4 | 464 | |||
GenBank mRNA | AK019704.1 | - | 3, 4 | 465 | ||
BY715476.1 | - | 3, 4 | 461 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV267612.1 | - | 1, 2 | 155 | Chr. 18 - 29452123 - 29466107 [+] on Build GRCm38 |
