siRNA Details
Gene Symbol: 2900073N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013778.1 | - | 1 | 41 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712745.1 | - | 1 | 39 | Chr. 2 - 17017810 - 17018644 [-] on Build GRCm38 |
