siRNA Details
Gene Symbol: C130023A14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047945.1 | - | 1 | 2645 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB364719.2 | - | 1 | 267 | Chr. 16 - 77648714 - 77649426 [+] on Build GRCm38 |
