siRNA Details
Gene Symbol: 2900006B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013488.1 | - | 1 | 1133 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV150459.1 | - | 1 | 70 | Chr. 11 - 90356562 - 90356738 [-] on Build GRCm38 |
