siRNA Details
Gene Symbol: 5830407E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017905.1 | - | 1 | 1117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DT929971.1 | - | 1 | 475 | Chr. 2 - 43995653 - 43996222 [+] on Build GRCm38 |
