siRNA Details
Gene Symbol: 1700010L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131796.1 | - | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB610327.1 | - | 1 | 172 | Chr. 19 - 56821962 - 56822539 [+] on Build GRCm38 |
