siRNA Details
Gene Symbol: 1700010B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005813.1 | - | 1 | 568 | ||
AK133430.1 | - | 1 | 548 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705966.1 | - | 1 | 550 | Chr. 15 - 73645869 - 73646515 [+] on Build GRCm38 |
