siRNA Details
Gene Symbol: 3110001L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013958.1 | - | 1 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712981.1 | - | 1 | 348 | Chr. 10 - 42089610 - 42090201 [+] on Build GRCm38 |
