siRNA Details
Gene Symbol: Stk38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020024.1 | - | 1 | 632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA526734.1 | - | 1 | 303 | Chr. 17 - 29006991 - 29007539 [-] on Build GRCm38 |
