siRNA Details
Gene Symbol: Gm29683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049871.1 | - | 3 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB598008.2 | - | 3 | 369 | Chr. 7 - 70273742 - 70337839 [-] on Build GRCm38 |
