siRNA Details
Gene Symbol: 1700018P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006100.1 | - | 1 | 81 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706212.1 | - | 1 | 81 | Chr. 9 - 8134513 - 8135844 [+] on Build GRCm38 |
