siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014591.1 | - | 1 | 2097 | ||
AK122552.1 | - | 11 | 4964 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014591.1 | - | 1 | 2097 | ||
AK122552.1 | - | 11 | 4964 |