siRNA Details
Gene Symbol: 4922502H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046187.1 | 3 | 261 | |||
GenBank mRNA | AK019555.1 | - | 3 | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY380565.1 | - | 3 | 239 | Chr. 13 - 101391871 - 101397558 [-] on Build GRCm38 |
