siRNA Details
Gene Symbol: 4921539H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019551.2 | - | 353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714555.1 | - | 2 | 353 | Chr. 3 - 58163932 - 58166532 [+] on Build GRCm38 |
