siRNA Details
Gene Symbol: 4930441J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040501.1 | 3 | 244 | |||
GenBank mRNA | AK015352.1 | - | 3 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714907.1 | - | 3 | 241 | Chr. 2 - 74298938 - 74307714 [-] on Build GRCm38 |
