siRNA Details
Gene Symbol: Gm6260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040405.1 | 1 | 132 | |||
GenBank mRNA | AK043613.1 | - | 1 | 134 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB641225.1 | - | 1 | 134 | Chr. 3 - 143671134 - 143712143 [-] on Build GRCm38 |
