siRNA Details
Gene Symbol: 2610300A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011940.1 | - | 1 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711143.1 | - | 1 | 365 | Chr. 1 - 38767184 - 38767688 [+] on Build GRCm38 |
