siRNA Details
Gene Symbol: 9430021M05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033569.1 | 4 | 1350 | |||
GenBank mRNA | AK079072.1 | - | 3 | 1275 | ||
AK163475.1 | - | 4 | 1202 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430021M05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033569.1 | 4 | 1350 | |||
GenBank mRNA | AK079072.1 | - | 3 | 1275 | ||
AK163475.1 | - | 4 | 1202 |