siRNA Details
Gene Symbol: 4930580E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016337.1 | - | 627 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715978.1 | - | 2 | 628 | Chr. 5 - 23387109 - 23388603 [+] on Build GRCm38 |
